Gene Transcripts
- Gene Symbol
- Gm10010
- Entrez Gene ID
- 434089
- Gene Name
- predicted gene 10010
- Gene Aliases
- ENSMUSG00000056771
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.101361
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039686.1 |
- |
2 |
910 |
|
View Full Product Details