Gene Transcripts
- Gene Symbol
- 1700066B17Rik
- Entrez Gene ID
- 73426
- Gene Name
- RIKEN cDNA 1700066B17 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 39842428 - 39847330 on Build GRCm38
- UniGene ID
- Mm.396239
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040465.1 |
|
5 |
426 |
|
GenBank mRNA |
AK006899.1 |
- |
3 |
158 |
|
BC048628.1 |
- |
5 |
426 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU962972.1 |
- |
5 |
429 |
|
Chr. 1 - 39842430 - 39847332 [-] |
View Full Product Details