Gene Transcripts
- Gene Symbol
- Plaa
- Entrez Gene ID
- 18786
- Gene Name
- phospholipase A2, activating protein
- Gene Aliases
- 2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3
- Chromosome Location
- Chr. 4: 94565139 - 94603247 on Build GRCm38
- UniGene ID
- Mm.490101
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085027.1 |
- |
1 |
684 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK164744.1 |
- |
|
692 |
|
Chr. - - [-] |
View Full Product Details