Gene Transcripts
- Gene Symbol
- Gm13093
- Entrez Gene ID
- 102641052
- Gene Name
- predicted gene 13093
- Gene Aliases
- OTTMUSG00000010397
- Chromosome Location
- Chr. 4: 150115742 - 150119287 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK081620.1 |
- |
|
2117 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB370270.2 |
- |
1 |
227 |
|
Chr. 4 - 150113881 - 150114474 [-] |
View Full Product Details