Gene Transcripts
- Gene Symbol
- 4933416E03Rik
- Entrez Gene ID
- 71081
- Gene Name
- RIKEN cDNA 4933416E03 gene
- Gene Aliases
- Chromosome Location
- Chr. 2: 159947341 - 159981289 on Build GRCm38
- UniGene ID
- Mm.383491
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040498.1 |
|
3 |
373 |
|
GenBank mRNA |
AK016829.1 |
- |
3 |
375 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY716503.1 |
- |
3 |
375 |
|
Chr. 2 - 159947341 - 159981092 [+] |
View Full Product Details