Gene Transcripts
- Gene Symbol
- 4930539C22Rik
- Entrez Gene ID
- 75208
- Gene Name
- RIKEN cDNA 4930539C22 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 134404143 - 134423635 on Build GRCm38
- UniGene ID
- Mm.84580
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040600.1 |
|
2 |
140 |
|
GenBank mRNA |
AK015994.1 |
- |
2 |
141 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715572.1 |
- |
|
141 |
|
Chr. - - [-] |
View Full Product Details