Gene Transcripts
- Gene Symbol
- C030006N10Rik
- Entrez Gene ID
- 78690
- Gene Name
- RIKEN cDNA C030006N10 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021051.1 |
- |
1 |
485 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY728112.1 |
- |
1 |
485 |
|
Chr. 10 - 32279159 - 32279693 [-] |
View Full Product Details