Gene Transcripts

Gene Symbol
Ints7
Entrez Gene ID
77065
Gene Name
integrator complex subunit 7
Gene Aliases
5930412E23Rik, int7
Chromosome Location
Chr. 1: 191575641 - 191622676 on Build GRCm38
UniGene ID
Mm.338175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020029.1 - 139
AK034856.1 - 20 4002
AK052103.1 - 18 3729
BC054120.1 - 20 3989 6506658

View Full Product Details