Gene Transcripts

Gene Symbol
Cyp11a1
Entrez Gene ID
13070
Gene Name
cytochrome P450, family 11, subfamily a, polypeptide 1
Gene Aliases
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc
Chromosome Location
Chr. 9: 58015017 - 58027023 on Build GRCm38
UniGene ID
Mm.302865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK042878.1 - 44
AK137397.1 - 17
AK163559.1 - 56


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ143316.1 - 1 52 Chr. 9 - 57998044 - 58005636 [+]
GenBankmRNA DN176714.1 - 1 25 Chr. 9 - 57998071 - 58005895 [+]
GenBankmRNA BY722569.1 - 1 17 Chr. 9 - 57998080 - 58005988 [+]
GenBankmRNA BY727381.1 - 1 52 Chr. 9 - 57998044 - 58005959 [+]
GenBankmRNA CJ143202.1 - 1 23 Chr. 9 - 57998074 - 58005730 [+]

View Full Product Details