Gene Transcripts
- Gene Symbol
- 1700067K01Rik
- Entrez Gene ID
- 73453
- Gene Name
- RIKEN cDNA 1700067K01 gene
- Gene Aliases
- Chromosome Location
- Chr. 8: 84000496 - 84004770 on Build GRCm38
- UniGene ID
- Mm.491533
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138628.1 |
- |
1 |
533 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY724382.1 |
- |
1 |
530 |
|
Chr. 8 - 84004235 - 84004775 [-] |
GenBankmRNA |
BB196432.1 |
- |
1 |
256 |
|
Chr. 8 - 84004235 - 84004490 [-] |
GenBankmRNA |
BM939124.1 |
- |
1 |
286 |
|
Chr. 8 - 84004152 - 84004529 [-] |
View Full Product Details