Gene Transcripts

Gene Symbol
PRKAR2A-AS1
Entrez Gene ID
100506637
Gene Name
PRKAR2A antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 3: 48847937 - 48851982 on Build GRCh38
UniGene ID
Hs.634259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_109996.1 2 632
NR_109997.1 3 553
GenBank mRNA AK127190.1 - 2 612
DA111905.1 - 3 554


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000412171.2 - 2 632 Chr. 3 - 48847937 - 48849454 [+]
GenBankmRNA DA244831.1 - 3 554 Chr. 3 - 48847937 - 48849175 [+]
GENCODE ENST00000416209.2 - 2 612 Chr. 3 - 48847957 - 48851981 [+]
GenBankmRNA DA190997.1 - 2 263 Chr. 3 - 48848306 - 48849454 [+]
GenBankmRNA DB082433.1 - 3 553 Chr. 3 - 48847937 - 48849167 [+]
GenBankmRNA DA237902.1 - 2 549 Chr. 3 - 48848020 - 48849190 [+]

View Full Product Details