Gene Transcripts

Gene Symbol
LINC01356
Entrez Gene ID
100996702
Gene Name
long intergenic non-protein coding RNA 1356
Gene Aliases
Chromosome Location
Chr. 1: 112820169 - 112850643 on Build GRCh38
UniGene ID
Hs.632431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103746.1 7 1577
GenBank mRNA AK093435.1 - 7 1577
BC019891.1 - 3 474
BC028023.1 - 3 468

View Full Product Details