Gene Transcripts
- Gene Symbol
- LOC101927532
- Entrez Gene ID
- 101927532
- Gene Name
- uncharacterized LOC101927532
- Gene Aliases
- Chromosome Location
- Chr. 1: 230002717 - 230007162 on Build GRCh38
- UniGene ID
- Hs.40183
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135116.1 |
|
2 |
300 |
|
GenBank mRNA |
DA255160.1 |
- |
2 |
300 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000445339.1 |
- |
2 |
287 |
|
Chr. 1 - 230002730 - 230007162 [+] |
GenBankmRNA |
H80797.1 |
- |
2 |
274 |
|
Chr. 1 - 230002743 - 230006881 [+] |
GenBankmRNA |
BQ188485.1 |
- |
2 |
288 |
|
Chr. 1 - 230002731 - 230007126 [+] |
View Full Product Details