Gene Transcripts
- Gene Symbol
- RALY-AS1
- Entrez Gene ID
- 101926888
- Gene Name
- RALY antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 20: 33992488 - 33994367 on Build GRCh38
- UniGene ID
- Hs.405849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109885.1 |
|
3 |
336 |
|
NR_109886.1 |
|
2 |
245 |
|
GenBank mRNA |
AA447916.1 |
- |
3 |
319 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000419662.2 |
- |
2 |
235 |
|
Chr. 20 - 33992511 - 33994357 [-] |
View Full Product Details