Gene Transcripts
- Gene Symbol
- LOC100506077
- Entrez Gene ID
- 100506077
- Gene Name
- uncharacterized LOC100506077
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.132801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX119135.1 |
- |
2 |
248 |
|
View Full Product Details