Gene Transcripts
- Gene Symbol
- LINC00448
- Entrez Gene ID
- 100874176
- Gene Name
- long intergenic non-protein coding RNA 448
- Gene Aliases
- Chromosome Location
- Chr. 13: 62672285 - 62807359 on Build GRCh38
- UniGene ID
- Hs.402083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047029.1 |
|
7 |
608 |
|
GenBank mRNA |
CR741874.1 |
- |
7 |
608 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000448411.1 |
- |
3 |
243 |
|
Chr. 13 - 62672285 - 62732180 [-] |
View Full Product Details