Gene Transcripts
- Gene Symbol
- NAALADL2-AS3
- Entrez Gene ID
- 100862679
- Gene Name
- NAALADL2 antisense RNA 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 175079312 - 175115242 on Build GRCh38
- UniGene ID
- Hs.148810
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046390.1 |
|
4 |
326 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000436929.1 |
- |
3 |
273 |
|
Chr. 3 - 175079312 - 175112600 [-] |
View Full Product Details