Gene Transcripts
- Gene Symbol
- FLJ22447
- Entrez Gene ID
- 400221
- Gene Name
- uncharacterized LOC400221
- Gene Aliases
- Chromosome Location
- Chr. 14: 61570540 - 61654713 on Build GRCh38
- UniGene ID
- Hs.344000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_039985.1 |
|
3 |
471 |
|
GenBank mRNA |
AK026100.1 |
- |
3 |
475 |
|
BQ439352.1 |
- |
3 |
564 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000229465.10 |
- |
3 |
471 |
|
Chr. 6 - 111308009 - 111315422 [-] |
GenBankmRNA |
BX339593.2 |
- |
3 |
414 |
|
Chr. 14 - 61570596 - 61653843 [+] |
GenBankmRNA |
BI830545.1 |
- |
3 |
392 |
|
Chr. 14 - 61647420 - 61654001 [+] |
GENCODE |
ENST00000556569.4 |
- |
3 |
415 |
|
Chr. 14 - 61570596 - 61654713 [+] |
GenBankmRNA |
BU933743.1 |
- |
3 |
347 |
|
Chr. 14 - 61570664 - 61654180 [+] |
View Full Product Details