Gene Transcripts
- Gene Symbol
- LOC403323
- Entrez Gene ID
- 403323
- Gene Name
- uncharacterized LOC403323
- Gene Aliases
- Chromosome Location
- Chr. 9: 62857856 - 62898087 on Build GRCh38
- UniGene ID
- Hs.646340
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_122076.1 |
|
5 |
1031 |
|
GenBank mRNA |
AK309896.1 |
- |
5 |
651 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000591993.5 |
- |
5 |
651 |
|
Chr. 9 - 62857833 - 62897707 [-] |
View Full Product Details