Gene Transcripts
- Gene Symbol
- LOC403323
- Entrez Gene ID
- 403323
- Gene Name
- uncharacterized LOC403323
- Gene Aliases
- Chromosome Location
- Chr. 9: 62857856 - 62898087 on Build GRCh38
- UniGene ID
- Hs.646340
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000612590.1 |
- |
|
630 |
|
Chr. - - [-] |
GenBankmRNA |
DA257209.1 |
- |
2 |
580 |
|
Chr. 9 - 62869224 - 62897932 [-] |
GenBankmRNA |
DC380731.1 |
- |
2 |
245 |
|
Chr. 9 - 62867024 - 62897707 [-] |
GenBankmRNA |
DC351384.1 |
- |
2 |
483 |
|
Chr. 9 - 62869190 - 62897835 [-] |
GENCODE |
ENST00000591993.5 |
- |
2 |
245 |
|
Chr. 9 - 62857833 - 62897707 [-] |
View Full Product Details