Gene Transcripts
- Gene Symbol
- LINC01239
- Entrez Gene ID
- 441389
- Gene Name
- long intergenic non-protein coding RNA 1239
- Gene Aliases
- Chromosome Location
- Chr. 9: 22646200 - 22824213 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038977.1 |
|
4 |
475 |
|
GenBank mRNA |
AK092601.1 |
- |
4 |
475 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000436786.1 |
- |
4 |
475 |
|
Chr. 9 - 22646200 - 22824213 [+] |
View Full Product Details