Gene Transcripts
- Gene Symbol
- RGMB-AS1
- Entrez Gene ID
- 503569
- Gene Name
- RGMB antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 98769618 - 98773084 on Build GRCh38
- UniGene ID
- Hs.586188
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000501938.6 |
- |
2 |
1938 |
|
Chr. 5 - 98769618 - 98773088 [-] |
GENCODE |
ENST00000498871.2 |
- |
3 |
1807 |
|
Chr. 5 - 98769618 - 98773125 [-] |
View Full Product Details