Gene Transcripts
- Gene Symbol
- LOC105377621
- Entrez Gene ID
- 105377621
- Gene Name
- uncharacterized LOC105377621
- Gene Aliases
- Chromosome Location
- Chr. 4: 102418602 - 102502486 on Build GRCh38
- UniGene ID
- Hs.15833
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136202.1 |
|
5 |
565 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000505709.1 |
- |
2 |
407 |
|
Chr. 4 - 102418602 - 102431282 [-] |
View Full Product Details