Gene Transcripts
- Gene Symbol
- LOC101928443
- Entrez Gene ID
- 101928443
- Gene Name
- uncharacterized LOC101928443
- Gene Aliases
- Chromosome Location
- Chr. 11: 70056230 - 70065371 on Build GRCh38
- UniGene ID
- Hs.313468
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120529.1 |
|
2 |
1629 |
|
NR_120530.1 |
|
2 |
422 |
|
GenBank mRNA |
BC039105.1 |
- |
2 |
1629 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000511013.2 |
- |
2 |
1629 |
|
Chr. 11 - 70056230 - 70065371 [-] |
View Full Product Details