Gene Transcripts
- Gene Symbol
- LOC101929153
- Entrez Gene ID
- 101929153
- Gene Name
- uncharacterized LOC101929153
- Gene Aliases
- Chromosome Location
- Chr. 5: 4773481 - 4774865 on Build GRCh38
- UniGene ID
- Hs.385526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104619.1 |
|
2 |
633 |
|
GenBank mRNA |
BC034630.1 |
- |
2 |
604 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000503757.1 |
- |
2 |
631 |
|
Chr. 5 - 4773481 - 4774865 [+] |
GenBankmRNA |
BG772454.1 |
- |
2 |
610 |
|
Chr. 5 - 4773508 - 4774853 [+] |
View Full Product Details