Gene Transcripts
- Gene Symbol
- LOC101929153
- Entrez Gene ID
- 101929153
- Gene Name
- uncharacterized LOC101929153
- Gene Aliases
- Chromosome Location
- Chr. 5: 4773481 - 4774865 on Build GRCh38
- UniGene ID
- Hs.385526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104619.1 |
|
1 |
34 |
|
GenBank mRNA |
BU852927.1 |
- |
1 |
35 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000503757.1 |
- |
1 |
32 |
|
Chr. 5 - 4773481 - 4774865 [+] |
View Full Product Details