Gene Transcripts
- Gene Symbol
- LINC01377
- Entrez Gene ID
- 102467074
- Gene Name
- long intergenic non-protein coding RNA 1377
- Gene Aliases
- Chromosome Location
- Chr. 5: 3177835 - 3181232 on Build GRCh38
- UniGene ID
- Hs.575250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104617.1 |
|
2 |
620 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000505396.2 |
- |
|
620 |
|
Chr. 5 - 3178095 - 3181232 [+] |
View Full Product Details