Gene Transcripts
- Gene Symbol
- LINC01182
- Entrez Gene ID
- 101929071
- Gene Name
- long intergenic non-protein coding RNA 1182
- Gene Aliases
- Chromosome Location
- Chr. 4: 13655179 - 13931228 on Build GRCh38
- UniGene ID
- Hs.543714
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_121681.1 |
|
4 |
1175 |
|
GenBank mRNA |
AK091889.1 |
- |
4 |
1172 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000510907.5 |
- |
4 |
1175 |
|
Chr. 4 - 13655179 - 13931228 [+] |
View Full Product Details