Gene Transcripts
- Gene Symbol
- LOC105369595
- Entrez Gene ID
- 105369595
- Gene Name
- uncharacterized LOC105369595
- Gene Aliases
- Chromosome Location
- Chr. 12: 585698 - 586486 on Build GRCh38
- UniGene ID
- Hs.504422
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134624.1 |
|
2 |
297 |
|
GenBank mRNA |
T68370.1 |
- |
2 |
298 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000538943.1 |
- |
2 |
297 |
|
Chr. 12 - 585865 - 586486 [-] |
View Full Product Details