Gene Transcripts
- Gene Symbol
- LOC101929164
- Entrez Gene ID
- 101929164
- Gene Name
- uncharacterized LOC101929164
- Gene Aliases
- Chromosome Location
- Chr. 19: 23015075 - 23023697 on Build GRCh38
- UniGene ID
- Hs.557249
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110746.1 |
|
2 |
529 |
|
GenBank mRNA |
AL133086.1 |
- |
2 |
529 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000565584.1 |
- |
2 |
529 |
|
Chr. 19 - 23015075 - 23023697 [-] |
View Full Product Details