Gene Transcripts

Gene Symbol
LINC01477
Entrez Gene ID
101927900
Gene Name
long intergenic non-protein coding RNA 1477
Gene Aliases
Chromosome Location
Chr. 18: 40066286 - 40099233 on Build GRCh38
UniGene ID
Hs.385522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110791.1 1 136
GenBank mRNA BC034625.1 - 1 136
BC045816.1 - 1 136


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000566101.1 - 1 136 Chr. 18 - 40066286 - 40099233 [+]

View Full Product Details