Gene Transcripts
- Gene Symbol
- LINC00909
- Entrez Gene ID
- 400657
- Gene Name
- long intergenic non-protein coding RNA 909
- Gene Aliases
- Chromosome Location
- Chr. 18: 74591774 - 74597835 on Build GRCh38
- UniGene ID
- Hs.61508
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000580048.1 |
- |
2 |
1755 |
|
Chr. 18 - 74591774 - 74597824 [-] |
GENCODE |
ENST00000577806.1 |
- |
2 |
670 |
|
Chr. 18 - 74593201 - 74597569 [-] |
View Full Product Details