Gene Transcripts
- Gene Symbol
- LINC00664
- Entrez Gene ID
- 400680
- Gene Name
- long intergenic non-protein coding RNA 664
- Gene Aliases
- Chromosome Location
- Chr. 19: 21483715 - 21503238 on Build GRCh38
- UniGene ID
- Hs.710554
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097381.1 |
- |
15 |
2893 |
|
BC040866.1 |
- |
2 |
896 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000596203.1 |
- |
2 |
896 |
|
Chr. 19 - 21494285 - 21499893 [+] |
View Full Product Details