Gene Transcripts
- Gene Symbol
- A930007I19Rik
- Entrez Gene ID
- 77779
- Gene Name
- RIKEN cDNA A930007I19 gene
- Gene Aliases
- Chromosome Location
- Chr. 19: 29503663 - 29521987 on Build GRCm38
- UniGene ID
- Mm.458738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132712.1 |
- |
|
727 |
|
- Gene Symbol
- 8430422M14Rik
- Entrez Gene ID
- 109319
- Gene Name
- RIKEN cDNA 8430422M14 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078814.1 |
- |
1 |
680 |
|
- Gene Symbol
- Gm16233
- Entrez Gene ID
- 100504497
- Gene Name
- predicted gene 16233
- Gene Aliases
- OTTMUSG00000031903
- Chromosome Location
- Chr. 3: 144947622 - 144953789 on Build GRCm38
- UniGene ID
- Mm.358802
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054018.1 |
- |
|
120 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB665739.1 |
- |
1 |
117 |
|
Chr. 3 - 144947563 - 144953892 [+] |
View Full Product Details