Gene Transcripts
- Gene Symbol
- Gm7271
- Entrez Gene ID
- 639545
- Gene Name
- predicted gene 7271
- Gene Aliases
- EG639545
- Chromosome Location
- Chr. 5: 76484077 - 76516628 on Build GRCm38
- UniGene ID
- Mm.478255
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033501.1 |
|
2 |
486 |
|
GenBank mRNA |
BC147734.1 |
- |
2 |
569 |
|
View Full Product Details