Gene Transcripts
- Gene Symbol
- 1700003C15Rik
- Entrez Gene ID
- 69345
- Gene Name
- RIKEN cDNA 1700003C15 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 11749833 - 11769308 on Build GRCm38
- UniGene ID
- Mm.425217
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045478.1 |
|
3 |
382 |
|
GenBank mRNA |
AK005625.1 |
- |
3 |
383 |
|
- Gene Symbol
- 4930437M23Rik
- Entrez Gene ID
- 73976
- Gene Name
- RIKEN cDNA 4930437M23 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 11485333 - 11501403 on Build GRCm38
- UniGene ID
- Mm.389073
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015330.1 |
- |
4 |
436 |
|
- Gene Symbol
- 4930558F17Rik
- Entrez Gene ID
- 75330
- Gene Name
- RIKEN cDNA 4930558F17 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 11109800 - 11126455 on Build GRCm38
- UniGene ID
- Mm.326776
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016172.1 |
- |
|
460 |
|
View Full Product Details