Gene Transcripts
- Gene Symbol
- Gm10421
- Entrez Gene ID
- 100038689
- Gene Name
- predicted gene 10421
- Gene Aliases
- ENSMUSG00000072792
- Chromosome Location
- Chr. 12: 117144523 - 117151269 on Build GRCm38
- UniGene ID
- Mm.445351
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033538.1 |
|
2 |
374 |
|
GenBank mRNA |
AK138376.1 |
- |
2 |
321 |
|
AK140057.1 |
- |
2 |
375 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM934951.1 |
- |
2 |
235 |
|
Chr. 12 - 117144569 - 117151129 [-] |
GenBankmRNA |
BY724059.1 |
- |
2 |
321 |
|
Chr. 12 - 117144739 - 117151215 [-] |
GenBankmRNA |
BB180191.2 |
- |
2 |
246 |
|
Chr. 12 - 117144530 - 117151138 [-] |
GenBankmRNA |
BY727040.1 |
- |
2 |
375 |
|
Chr. 12 - 117147824 - 117151269 [-] |
View Full Product Details