Gene Transcripts
- Gene Symbol
- 1700072B07Rik
- Entrez Gene ID
- 73529
- Gene Name
- RIKEN cDNA 1700072B07 gene
- Gene Aliases
- Chromosome Location
- Chr. 9: 58370504 - 58374183 on Build GRCm38
- UniGene ID
- Mm.490924
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040727.1 |
|
3 |
218 |
|
GenBank mRNA |
AK006934.1 |
- |
3 |
218 |
|
View Full Product Details