Gene Transcripts
- Gene Symbol
- Gm29128
- Entrez Gene ID
- 102634543
- Gene Name
- predicted gene 29128
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.387028
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139037.1 |
- |
2 |
641 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY724809.1 |
- |
2 |
641 |
|
Chr. 1 - 31117139 - 31136638 [+] |
View Full Product Details