Gene Transcripts
- Gene Symbol
- 1700029M03Rik
- Entrez Gene ID
- 69476
- Gene Name
- RIKEN cDNA 1700029M03 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 164960664 - 164961400 on Build GRCm38
- UniGene ID
- Mm.67177
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006507.1 |
- |
2 |
233 |
|
AV045468.2 |
- |
1 |
184 |
|
AV205680.1 |
- |
2 |
190 |
|
View Full Product Details