Gene Transcripts
- Gene Symbol
- 4933400F21Rik
- Entrez Gene ID
- 74403
- Gene Name
- RIKEN cDNA 4933400F21 gene
- Gene Aliases
- 1700091C04Rik, 4930565K10Rik
- Chromosome Location
- Chr. 1: 89676507 - 89683892 on Build GRCm38
- UniGene ID
- Mm.444291
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715838.1 |
- |
1 |
200 |
|
Chr. 1 - 89650641 - 89683908 [-] |
GenBankmRNA |
BB616064.1 |
- |
1 |
164 |
|
Chr. 1 - 89680815 - 89683870 [-] |
GenBankmRNA |
CJ047535.1 |
- |
1 |
190 |
|
Chr. 1 - 89681033 - 89683898 [-] |
GenBankmRNA |
BB615358.1 |
- |
1 |
184 |
|
Chr. 1 - 89679411 - 89683892 [-] |
GenBankmRNA |
BB616490.1 |
- |
1 |
176 |
|
Chr. 1 - 89680820 - 89683883 [-] |
View Full Product Details