Gene Transcripts
- Gene Symbol
- LINC00115
- Entrez Gene ID
- 79854
- Gene Name
- long intergenic non-protein coding RNA 115
- Gene Aliases
- NCRNA00115
- Chromosome Location
- Chr. 1: 826206 - 827522 on Build GRCh38
- UniGene ID
- Hs.351445
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024321.1 |
|
1 |
1199 |
|
GenBank mRNA |
AK026292.1 |
- |
1 |
1184 |
|
BC017762.1 |
- |
1 |
1200 |
|
View Full Product Details