Gene Transcripts
- Gene Symbol
- NUTM2B-AS1
- Entrez Gene ID
- 101060691
- Gene Name
- NUTM2B antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 79691500 - 79826594 on Build GRCh38
- UniGene ID
- Hs.440648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120611.1 |
|
6 |
4253 |
|
GenBank mRNA |
AK091744.1 |
- |
1 |
2429 |
|
View Full Product Details