Gene Transcripts

Gene Symbol
LINC00909
Entrez Gene ID
400657
Gene Name
long intergenic non-protein coding RNA 909
Gene Aliases
Chromosome Location
Chr. 18: 74591774 - 74597835 on Build GRCh38
UniGene ID
Hs.61508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024484.1 2 1693
GenBank mRNA AK001853.1 - 2 1440
BC036588.2 - 2 1684
BC132930.1 - 2 1469
BC132956.1 - 2 1469

View Full Product Details