Gene Transcripts
- Gene Symbol
- LOC100506675
- Entrez Gene ID
- 100506675
- Gene Name
- uncharacterized LOC100506675
- Gene Aliases
- Chromosome Location
- Chr. 11: 32035979 - 32041318 on Build GRCh38
- UniGene ID
- Hs.446169
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX114263.1 |
- |
1 |
37 |
|
Chr. 11 - 32036718 - 32041268 [-] |
GenBankmRNA |
DB024222.1 |
- |
1 |
60 |
|
Chr. 11 - 32040000 - 32041291 [-] |
GenBankmRNA |
AA301105.1 |
- |
1 |
46 |
|
Chr. 11 - 32036893 - 32041277 [-] |
View Full Product Details