Gene Transcripts
- Gene Symbol
- LOC101927168
- Entrez Gene ID
- 101927168
- Gene Name
- uncharacterized LOC101927168
- Gene Aliases
- Chromosome Location
- Chr. 18: 6728927 - 6729862 on Build GRCh38
- UniGene ID
- Hs.242240
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134645.1 |
|
2 |
356 |
|
GenBank mRNA |
BG202214.1 |
- |
1 |
217 |
|
BX097886.1 |
- |
2 |
356 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB335425.1 |
- |
1 |
26 |
|
Chr. 18 - 6728475 - 6728981 [-] |
View Full Product Details