Gene Transcripts

Gene Symbol
LINC01518
Entrez Gene ID
101929397
Gene Name
long intergenic non-protein coding RNA 1518
Gene Aliases
Chromosome Location
Chr. 10: 42673013 - 42691759 on Build GRCh38
UniGene ID
Hs.729796
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120656.1 5 627
NR_120657.1 4 521
NR_120658.1 4 517
NR_120659.1 5 638
GenBank mRNA AK123067.1 - 5 627

View Full Product Details