Gene Transcripts

Gene Symbol
SERHL
Entrez Gene ID
94009
Gene Name
serine hydrolase-like (pseudogene)
Gene Aliases
BK126B4.1, BK126B4.2, HS126B42, dJ222E13.1
Chromosome Location
Chr. 22: 42500579 - 42512560 on Build GRCh38
UniGene ID
Hs.360940
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027786.1 1 90
GenBank mRNA AK094872.1 - 1 90
AL365513.1 - 1 79
BC093888.1 - 1 44 7939733
DA273263.1 - 1 90

View Full Product Details