Gene Transcripts
- Gene Symbol
- LOC55338
- Entrez Gene ID
- 55338
- Gene Name
- uncharacterized LOC55338
- Gene Aliases
- Chromosome Location
- Chr. 5: 87215735 - 87239201 on Build GRCh38
- UniGene ID
- Hs.677194
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AU134573.1 |
- |
2 |
173 |
|
Chr. 5 - 87217892 - 87239201 [-] |
GenBankmRNA |
AU137242.2 |
- |
1 |
36 |
|
Chr. 5 - 87217805 - 87238981 [-] |
View Full Product Details