Gene Transcripts
- Gene Symbol
- LOC101928580
- Entrez Gene ID
- 101928580
- Gene Name
- uncharacterized LOC101928580
- Gene Aliases
- Chromosome Location
- Chr. 11: 74311362 - 74324705 on Build GRCh38
- UniGene ID
- Hs.569025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120556.1 |
|
5 |
1044 |
|
GenBank mRNA |
BC039366.1 |
- |
5 |
1042 |
|
BC048427.1 |
- |
5 |
1042 |
|
View Full Product Details